Professional Washing Machines, PWM
Supplier: Miele
11803110 11803300 11803070
76936-192EA 29866.01 CAD
76936-192 76936-210 76936-186
Professional Washing Machines, PWM
Washing Machines and Tumble Dryers
Professional washing machines, electric heating with detergent drawer, M Touch Pro Plus – freely programmable.
  • M Touch Pro Plus freely programmable color display
  • Short program cycles
  • Honeycomb drum 2.0
  • 3D imbalance monitoring
  • G-force of 458 (PWM 912) or 462 (PWM 916 and PWM 920)
  • High durability: tested for 30000 operating hours
  • Safe disinfection
  • Low consumption values
Honeycomb drum 2.0: the scoop rib perforations in the honeycomb drum 2.0 cause water to shower down on the load, guaranteeing thorough soaking.
Short crogram cycles: premiumMore Product Information