MULTISCREEN™ Human Glucagon CHO-K1 HTS Membranes
Supplier: MULTISPAN INC
MC1266-1-100MG MC1266-1-10MG MC1266-1-1MG MC1266-1-5MG MC1266-1A100MG MC1266-1A-10MG MC1266-1A-1MG MC1266-1A-5MG
76921-350EA 49827.7 CAD
76921-350 76923-274 76922-248 76922-042 76923-682 76923-542 76920-052 76921-772
MULTISCREEN™ Human Glucagon CHO-K1 HTS Membranes
Cells
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage conditions LN₂ (Below –135 ℃)