Anti-PKN1 Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A12648-100
76848-678EA 555.69 CAD
76848-678 76862-920
Anti-PKN1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PKN1 for WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:100

Type: Primary
Antigen: PKN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

SPECIFICATIONS

Antigen symbol PKN1
Environmentally Preferable
Antigen name DBK
Antigen synonyms PKN|PKN ALPHA|PKC1|PAK 1|PKN1|PAK-1|PAK1|PRK1|PKN1_HUMAN
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID UniprotID# Q16512
Isotype IgG
Western blot Yes
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human PKN1 (NP_002732.3)
Sequence MASDAVQSEPRSWSLLEQLGLAGADLAAPGVQQQLELERERLRREIRKELKLKEGAENLRRATTDLGRSLGPVELLLRGSSRRLDLLHQQLQELHAHVVLPDPAATHDGPQSPGAGGPTCSATNLSRVAGLEKQLAIELKVKQGAENMIQTYSNGSTKDRKLLLTAQQMLQDSKTKIDIIRMQLRRALQAGQLENQAAPDDTQGSPDLGAVELRIEELRHHFRVEHAVAEGAKNVLRLLSAAKAPDRKAVSEAQEKLTESNQKLGLLREALERRLGELPADHPKGRLLREELAAASSAAF
Form Liquid
Molecular weight 120 kDa
Purification Affinity purification.
Storage buffer Supplied in Phosphate buffered saline, pH 7.3, with 50% Glycerol and 0.02% Sodium Azide
Storage temperature Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
Shipping temperature Shipped on blue ice at +4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR