Rental Sampling Kits
Supplier: STAT PEEL INC
A00555 A00556 A00557
76848-034EA 10520.52 CAD
76848-034 76848-044 76848-038
Rental Sampling Kits
Samplers Air Samplers
Sampling kits allow for the collection of respirable, inhalable, or both respirable and inhalable particles for inspection in the STAT PEEL® Identifier System.
  • Battery powered badges for particle collection
  • Tablet computer for badge control
  • Badges chargers
  • All necessary tools to perform air samplings
  • Starter kit of 20 filtration slides
The sampling kit allows for the collection of respirable, inhalable, or both respirable and inhalable particles for inspection in the STAT PEEL® Identifier system or for SEM inspection. The included filtration badgesMore Product Information