Vessel Packages for HABITAT research Bioreactors
Supplier: IKA Works
10007659 10007660 10007661 10007662 10007646 10007647 10007648 10007649 10007650 10007651 10007652 10007655 10007657 10007658 10007644 10007645
76836-722EA 21499.72 CAD
76836-722 76836-706 76836-680 76836-694 76836-690 76836-686 76836-698 76836-684 76836-672 76836-716 76836-676 76836-708 76836-710 76836-678 76836-724 76836-688
Vessel Packages for HABITAT research Bioreactors
Bioreactors
Delivery information: Cell vessels are supplied with: Glass vessel (single or double walled), vessel holder, lid complete, driving shaft, motor coupling, micro sparger, condenser, three pitch blade impeller, temp. sensor, pH sensor, DO sensor, foam sensor, level sensor, heating blanket (for sw package), cable and tube set, 4× sample flask 250 ml and syringe set.
Ferment vessels are supplied with: Glass vessel (single or double walled), vessel holder, lid complete, driving shaft, motor coupling, ring sparger, condenser, baffle, six blade segment impeller, temp. sensor, pH sensor, DO sensor,More Product Information