Suite® Touchless Primary Duty Mop Pad, Benchmark Products
Supplier: BENCHMARK PRODUCTS INC. TE
Y-TL-517-25
76836-666CS 560.9 CAD
76836-666
Suite® Touchless Primary Duty Mop Pad, Benchmark Products
Mops
Suite® touchless primary duty mops eliminate operator glove handling of both clean and sterile or dirty pads. It provides consistent and liberal application of disinfectants.
The mop pad incorporates Suite's engineered fiber resulting in more square feet of application per prepped pad. Additionally, the touch-less design also significantly reduces time spent changing pads.
Ordering information: Handle, frame and strips have to be ordered separately.
Packaging: 5 mop pads per pack, 25 mops per case.