Corepoint® Scientific Full Size Sliding Wire Basket for Select Refrigerator Units
Supplier: Horizon Scientific
CPS-BSKT-BASE-FIXED-0504 CPS-BSKT-BASE-FIXED-10.5 CPS-SLD-BSKT-FULL-0504 CPS-SLD-BSKT-FULL-10.5
76809-174EA 223.83 CAD
76809-174 76809-176 76809-178 76809-180
Corepoint® Scientific Full Size Sliding Wire Basket for Select Refrigerator Units
Baskets Wire Baskets
Corepoint® Scientific sliding storage baskets can streamline your inventory management and enhance the safety and security of your products.
  • Enhanced organization
  • Improved air circulation
  • Maximized storage space
Baskets are designed to fit 10.5 cu. ft. compact refrigerators and 5.2 cu. ft. freestanding undercounter refrigerators. Their sturdy construction allows you to stack and organize inventory for efficiency and easy accessibility.
Baskets are designed to promote superior air circulation, preventing hot spots and maintaining consistentMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable