Freezer, Full Stainless Door with Lock
Supplier: Nordon/Hoshizaki Llc.
F1A-FS F2A-FS
76808-994EA 18859.21 CAD
76808-994 76808-996
Freezer, Full Stainless Door with Lock
Freezers Laboratory / General Purpose Freezers
The Steelheart series upright single and two section freezer with full stainless doors is an attractive workhorse for any business. The dependable 1 and ³/₄ HP motor that powers the compressor in the top of this unit provides the cabinet with a continuous stream of cold air through its unique ‘ducted’ air distribution system, which ensures a steady constant temperature.
  • Easy to clean
  • Lockable full stainless door
  • F1A-FS Includes 3 epoxy coated wire
  • F2A-FS Includes 6 epoxy coated wire
  • Epoxy electrocoated (E-Coat) evaporator coil to help fight corrosion for
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Material Stainless steel Stainless steel
Weight 161 kg (355 lbs.) 254 kg (560 lbs.)