Hot Plate Stirrers, AREX 5
Supplier: VELP SCIENTIFIC INC.
F20510570 SA20510570
76795-554EA 1061.51 CAD
76795-554 76795-560
Hot Plate Stirrers, AREX 5
Stirrers Magnetic Hotplate Stirrers
The AREX 5 is a robust and versatile hotplate stirrer suitable for applications ranging from basic to temperature-sensitive where precise sample thermoregulation is required.
  • Easy-to-use and safe
  • Ensures a high level of safety and protection for laboratory operators
  • Two illuminated indicators always inform the operator when the heating is on
  • The 135 mm diameter top is protected from chemicals and scratches by the superior resistant CerAlTop™ ceramic coating
The AREX 5 can work stand-alone or be upgraded to digital by connecting the VTF digitalMore Product Information