Sterilization Wraps, 2-Ply SMS, One-Step Heavy Duty
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
KCRP1-300-4040 KCRP1-300-4545 KCRP1-300-4548 KCRP1-300-4848 KCRP1-300-5454 KCRP1-300-5472 KCRP1-300-0808 KCRP1-300-0909 KCRP1-300-0912 KCRP1-300-1212 KCRP1-300-1515 KCRP1-300-1818 KCRP1-300-2020 KCRP1-300-2424 KCRP1-300-3030 KCRP1-300-3636 KCRP1-300-5490 KCRP1-300-6060 KCRP1-300-6080
76795-492CS 1238.33 CAD
76795-492 76795-480 76795-472 76795-496 76795-474 76795-476 76795-470 76795-494 76795-486 76795-488 76795-498 76795-466 76795-482 76795-468 76795-464 76795-490 76795-484 76795-478 76795-462
Sterilization Wraps, 2-Ply SMS, One-Step Heavy Duty
Sterilization Wraps
One-Step super duty SMS 2-ply wrappers provide an excellent microbial barrier. Ideal for pharmaceutical production applications.
  • Manufactured and packaged in a certified ISO 8 Cleanroom
  • 2-ply
  • Clean - no cellulose
  • Strong, rip and tear resistant 88 gsm SMS material
  • An extensive range of sizes available means less cutting
  • Hydrophobic barrier allows sterilization gases, but prevents liquid and microbial penetration
  • Gram weight: 50 (each layer)
For use in steam, ethylene oxide and hydrogen peroxide sterilization processes.
Delivery information: More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable