Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Supplier: AAT BIOQUEST INC
22327
76779-086EA 880.2 CAD
76779-086
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Assays Cellular Assays
Provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry-based assay
  • alternative to anti-BrdU
  • extremely sensitive
  • red fluorescence
The resulted iFluor® 555-labeled DNA formed in cells is imaged with a Cy3 filter set.
Uses XdU which is incorporated into cellular DNA during DNA synthesis