Rat GSTM2 ELISA Kit

Supplier: ANTIBODIES.COM LLC

A303756-96
76747-396EA 1240.14 CAD
76747-396
Rat GSTM2 ELISA Kit
Assays ELISAs

Rat GSTM2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat GSTM2 in serum, plasma, tissue homogenates, and other biological fluids.


  • Ready-to-use ELISA kit
  • Detection range: 0.156 to 10 ng/ml
  • Sensitivity: 0.094 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%


Rat GSTM2 ELISA kit (A303756) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat GSTM2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for GSTM2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the GSTM2 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-GSTM2 antibody, which binds the captured GSTM2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of GSTM2 captured in each well. The concentration of GSTM2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Ordering information: This is a ready-to-use ELISA kit that contains: pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR