Human Muscarinic Acetylcholine Receptor ELISA Kit

Supplier: ANTIBODIES.COM LLC

A74884-96
76740-474EA 1006.62 CAD
76740-474
Human Muscarinic Acetylcholine Receptor ELISA Kit
Assays ELISAs

Human Muscarinic Acetylcholine Receptor ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Muscarinic Acetylcholine Receptor in serum, plasma, tissue homogenates, and other biological fluids.


  • Ready-To-Use ELISA Kit
  • Detection Range: 0.625 - 40 ng/ml
  • Sensitivity: 0.375 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%


Human Muscarinic Acetylcholine Receptor ELISA kit (A74884) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Muscarinic Acetylcholine Receptor in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Muscarinic Acetylcholine Receptor has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Muscarinic Acetylcholine Receptor present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Muscarinic Acetylcholine Receptor Antibody, which binds the captured Muscarinic Acetylcholine Receptor present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Muscarinic Acetylcholine Receptor captured in each well. The concentration of Muscarinic Acetylcholine Receptor can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR