Virus Recombinant SARS-CoV-2 Omicron BA.2.75 Spike Protein Receptor Binding Domain (RBD) (from Insect cell), His Tag
Supplier: ReVacc Scientific
Z16Z77-100UG Z16Z77-500UG
76729-542EA 606.58 CAD
76729-542 76729-580
Virus Recombinant SARS-CoV-2 Omicron BA.2.75 Spike Protein Receptor Binding Domain (RBD) (from Insect cell), His Tag
Proteins and Peptides
A recombinant form of the receptor binding domain (RBD) with amino acids 319 to 541) of the spike (S) glycoprotein gene from SARS-CoV-2 bearing mutations identified in variant of Omicron BA.2.75, was produced by insect cells, followed by purification.
  • This product contains C-terminal 6 hexa-histidine tag
This product has been run on SDS-PAGE with reduced condition. We also evaluated its binding to ACE2-Fc, anti-His mAb and a negative mAb in ELISAs.