Keyguard® Sleeves
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
AG-KG
76717-458CS 114.91 CAD
76717-458
Keyguard® Sleeves
Disposable Garments
An economical alternative to Tyvek PPE.
  • 18×9" sleeve
  • Non-linting
  • Disposable
Breathable, non-linting sleeves are packaged 100 pairs per case.