Corepoint® Scientific Flammable Material Storage Refrigerators
Supplier: Horizon Scientific
FR141WWW/0HC FR171WWW/0HC FR201WWW/0HC
76717-368EA 7906.46 CAD
76717-368 76717-372 76717-376
Corepoint® Scientific Flammable Material Storage Refrigerators
Refrigerators
Safeguard volatile substances in these specially-designed flammable storage refrigerators.
  • Laboratory use
  • Microprocessor temperature controller
  • Temperature alarms
  • Keyed door locks
  • Probe access
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These flammable storage refrigerators meet the requirements of NFPA 45 and NFPA 30 for the cold storage of flammable and combustible materials in unclassified laboratory locations. They utilize non-sparking and/or ATEX rated components to ensure an intrinsically safe interior for flammable materials inMore Product Information