Corepoint® Scientific Hazardous Location Laboratory Refrigerators
Supplier: Horizon Scientific
ER141WWW/0HC ER171WWW/0HC ER201WWW/0HC
76717-354EA 10777.95 CAD
76717-354 76717-356 76717-358
Corepoint® Scientific Hazardous Location Laboratory Refrigerators
Refrigerators
These refrigerators are designed to keep you and your stored substances safe in work areas where hazardous conditions may exist.
  • Laboratory use
  • Microprocessor temperature controller
  • Temperature alarms
  • Keyed door locks
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These units utilize non-sparking and/or ATEX rated components and are engineered for use in environments where volatile/explosive conditions could potentially exist.
Models feature a microprocessor temperature controller, allowing for precise temperature control, recovery andMore Product Information