MyPIPETMAN® Select Pipettors
Supplier: GILSON, INC.
FP10011S FP10012S FP10013S FP10014S FP10015S FP10016S FP10001S FP10002S FP10003S FP10004S FP10005S FP10006S FP10007S FP10008S FP10009S FP10010S
76716-740EA 2150.1 CAD
76716-740 76716-742 76716-744 76716-746 76716-748 76716-750 76716-720 76716-722 76716-724 76716-726 76716-728 76716-730 76716-732 76716-734 76716-736 76716-738
MyPIPETMAN® Select Pipettors
Pipettes Micropipettes
Ergonomic, safe and reliable, MyPIPETMAN® Select offers volume setting choice and optional handle personalization for optimal use in various protocols.
  • Variable volume single channel models covering a volume range from 0.2 μl to 10 ml
  • Variable volume multichannel models covering a volume range from 0.5 μl to 300 μl (8 and 12-channel models)
  • Free volume setting position for quick and effortless volume adjustment
  • Light ratchet volume setting for precise volume adjustment
  • Volume locked position to secure pipetting
Set and secure pipetting volumes to meetMore Product Information