Mouse Leucine-rich Repeat LGI Family Member 2(LGI2) ELISA Kit

Supplier: AFG Bioscience

EK731215
76710-478EA 601.32 CAD
76710-478
Mouse Leucine-rich Repeat LGI Family Member 2(LGI2) ELISA Kit
Assays ELISAs

  • Detection wavelength: 450 nm

Order Now

SPECIFICATIONS

Environmentally Preferable
Target protein Leucine-rich repeat LGI family member 2
Primary antibody reactivity
Assay Type Sandwich
Host Mouse
Sample type Serum, plasma, urine, tissue homogenates, cell culture supernates
Sample volume 50 - 100 μl
Time to Results 1 - 5 h
Regulatory status RUO
Storage temperature 2…8 °C
Shelf life 6 months (4 °C); 12 months (−20 °C)
Application Neuroscience

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR