Human 1,25- Hydroxy Vitamin D3 (1,25(OH) 2 D3) ELISA Kit

Supplier: AFG Bioscience

EK710157
76709-002EA 601.32 CAD
76709-002
Human 1,25- Hydroxy Vitamin D3 (1,25(OH) 2 D3) ELISA Kit
Assays ELISAs

  • Detection wavelength: 450 nm

Order Now

SPECIFICATIONS

Target protein 1,25- hydroxy vitamin D3
Environmentally Preferable
Primary antibody reactivity
Assay Type Sandwich
Assay Principle Quantitative
Host Human
Sensitivity 2 μg/L
Detection range 13.5 - 100 μg/L
Sample type Serum, plasma, urine, tissue homogenates, cell culture supernates
Sample volume 50 - 100 µl
Time to Results 1 - 5 h
Regulatory status RUO
Storage temperature 2…8 °C
Shelf life 6 months (4 °C); 12 months (–20 °C)
Application Metabolism

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR