Human Complement 1q, C1q ELISA Kit

Supplier: AFG Bioscience

EK714354
76707-802EA 601.32 CAD
76707-802
Human Complement 1q, C1q ELISA Kit
Assays ELISAs

  • Detection wavelength: 450 nm

Order Now

SPECIFICATIONS

Target protein Complement 1q,C1q
Environmentally Preferable
Primary antibody reactivity
Assay Type Sandwich
Assay Principle Quantitative
Host Human
Sensitivity 0.039 ng/ml
Detection range 0.375 - 12 ng/ml
Sample type Serum, plasma, urine, tissue homogenates, cell culture supernates
Sample volume 50 - 100 µl
Time to Results 1 - 5 h
Regulatory status RUO
Storage temperature 2…8 °C
Shelf life 6 months (4 °C); 12 months (−20 °C)
Application Immunology

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR