Human Interferon-Inducible Protein 10(IP-10) ELISA Kit

Supplier: AFG Bioscience

EK710833
76706-252EA 601.32 CAD
76706-252
Human Interferon-Inducible Protein 10(IP-10) ELISA Kit
Assays ELISAs

  • Detection wavelength: 450 nm

Order Now

SPECIFICATIONS

Target protein Interferon-inducible protein 10
Environmentally Preferable
Primary antibody reactivity
Assay Type Sandwich
Assay Principle Quantitative
Host Human
Sensitivity 1 ng/L
Detection range 5 - 100 ng/L
Sample type Serum, plasma, urine, tissue homogenates, cell culture supernates
Sample volume 50 - 100 µl
Time to Results 1 - 5 h
Regulatory status RUO
Storage temperature 2…8 °C
Shelf life 6 months (4 °C); 12 months (–20 °C)
Application Immunology

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR