Human Total Iron Binding Capacity(TIBC) ELISA Kit

Supplier: AFG Bioscience

EK710467
76702-994EA 601.32 CAD
76702-994
Human Total Iron Binding Capacity(TIBC) ELISA Kit
Assays ELISAs

  • Detection wavelength: 450 nm

Order Now

SPECIFICATIONS

Target protein Total Iron binding capacity
Environmentally Preferable
Primary antibody reactivity
Assay Type Sandwich
Assay Principle Quantitative
Host Human
Sensitivity 0.5 nmol/ml
Detection range 2 - 85nmol/ml
Sample type Serum, plasma, urine, tissue homogenates, cell culture supernates
Sample volume 50 - 100 µl
Time to Results 1 - 5 h
Regulatory status RUO
Storage temperature 2…8 °C
Shelf life 6 months (4 °C); 12 months (–20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR