Human Cardiac Ankyrin Repeat Domain 1 (ANKRD1) ELISA Kit

Supplier: AFG Bioscience

EK714708
76699-876EA 601.32 CAD
76699-876
Human Cardiac Ankyrin Repeat Domain 1 (ANKRD1) ELISA Kit
Assays ELISAs

  • Detection wavelength: 450 nm

Order Now

SPECIFICATIONS

Environmentally Preferable
Target protein Cardiac ankyrin repeat domain 1
Primary antibody reactivity
Assay Type Sandwich
Assay Principle Quantitative
Host Human
Sensitivity 0.78 pg/ml
Detection range 6.25 - 200 pg/ml
Sample type Serum, plasma, urine, tissue homogenates, cell culture supernates
Sample volume 50 - 100 µl
Time to Results 1 - 5 h
Regulatory status RUO
Storage temperature 2…8 °C
Shelf life 6 months (4 °C); 12 months (−20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR