Conditioned Place Preference (CPP), Spatial with Corner Inserts
Supplier: MAZE ENGINEERS
4401 4402 4403 4404 4421 4422
76670-026EA 3981.6 CAD
76670-026 76670-028 76670-030 76670-032 76670-042 76670-044
Conditioned Place Preference (CPP), Spatial with Corner Inserts
Animal Supplies Animal Environmental Enrichment
The conditioned place preference chamber is a paradigm widely used to explore the reinforcing effects of natural and stimuli. The spatial conditioned place preference chamber is a modification of the original CPP chamber that assesses the similar paradigms as the standard CPP chamber, but includes standing inserts for spatial place preference procedures.
  • Acrylic construction
  • Easy to clean
  • Odor-free
  • Customizable colors for walls, floors, and corners also available
In this variant, subjects are allowed to freely move between a compartment in which theyMore Product Information