Morris Water Mazes
Supplier: MAZE ENGINEERS
3103 3101
76669-846EA 5240.35 CAD
76669-846 76669-844
Morris Water Mazes
Animal Supplies Animal Cages
The Morris water maze (MWM) and additional accessories are used to assess spatial learning and memory in rodents.
  • Made of plastic, and is availble in white and blue
  • An adjustable platform is included with every MWM order
The maze consists of a large circular pool filled with water and a hidden platform. The subject is placed in the pool and is required to find the hidden platform in order to escape the water.
Accessories information: Optional steel frame with casters is designed to fit the MWM with side drain. The optional radial arm inserts, T-More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable