Alsto JC5030 Jaw Crusher
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
402249 402250
76638-298EA 474408.76 CAD
76638-298 76638-300
Alsto JC5030 Jaw Crusher
Mills
The Alsto JC5030 is the largest jaw crusher. A high capacity unit suitable for laboratory use and plant applications. Rugged construction and high reduction capabilities together with low maintenance requirements make it outstanding for a large variety of applications.
  • Large input particle size capability
  • 500×300 mm jaw opening allows for lump sizes of up to 250 mm
  • 30 mm product size allows for fast, efficient reduction with a high throughput of 15,000 kg/hr
  • Simple and effective single toggle design reduces maintenance and repair time
  • Fast and easy jaw gap
More Product Information