ABS® Upright Refrigerators, Certified to the NSF/ANSI 456 Standard for Vaccine Storage, Horizon Scientific
Supplier: Horizon Scientific
PH-ABT-NSF-10PG PH-ABT-NSF-10PS PH-ABT-NSF-12G PH-ABT-NSF-12S PH-ABT-NSF-16G PH-ABT-NSF-16S PH-ABT-NSF-26G PH-ABT-NSF-26S PH-ABT-NSF-49G PH-ABT-NSF-49S PH-ABT-NSF-S12G PH-ABT-NSF-S12S PH-ABT-NSF-S16G PH-ABT-NSF-S16S PH-ABT-NSF-S26G PH-ABT-NSF-S26S PH-ABT-NSF-S49G PH-ABT-NSF-S49S
76637-278EA 9330.2 CAD
76637-278 76637-302 76637-280 76637-304 76637-282 76637-306 76637-284 76637-308 76637-286 76637-310 76637-288 76637-312 76637-290 76637-314 76637-292 76637-316 76637-294 76637-318
ABS® Upright Refrigerators, Certified to the NSF/ANSI 456 Standard for Vaccine Storage, Horizon Scientific
Refrigerators
These laboratory and pharmaceutical refrigerators offer premium protection for vaccine storage.
  • Microprocessor controller with LED display
  • Integrated audible and visual alarms
  • Hermetic, high performance compressor
  • White powder coated steel construction
  • Natural hydrocarbon refrigerants
  • NSF/ANSI 456 Vaccine Storage Certification
  • ENERGY STAR certified*
These premier refrigerators are designed in accordance with the NSF/ANSI 456 Standard for Vaccine Storage. Units protect pharmaceuticals at optimal temperatures, preventing waste and allowing for peakMore Product Information