PTFE FTIR Gas Cell, Type TGC-10, Lightpath 100 mm, Firebird Optics
Supplier: FIREBIRD OPTICS
TGC-10-BAF2 TGC-10-KCL TGC-10-NACL TGC-10-ZNSE TFC-10-CAF2 TGC-10
76636-408EA 4230.56 CAD
76636-408 76636-412 76636-414 76636-416 76636-404 76636-406
PTFE FTIR Gas Cell, Type TGC-10, Lightpath 100 mm, Firebird Optics
Optical Windows
The TGC-10 is a cell where all components of the cell that contacts the sample are made from PTFE. This is helpful for any sample that could corrode or be corroded by optical glass, Fused silica or stainless steel. This type of cell is most helpful in qualitative analysis applications where one or more gasses are present in concentrations of more than 5%.
  • Gas cell is constructed in PTFE
  • O-rings are used each end to seal windows with anodized aluminum caps
  • Two PFA valves are connected to body of gas cell
  • Window size 38 Ø × 6 mm, clear aperture 25 mm
More Product Information