InTemp® CX Gateway
Supplier: ONSET COMPUTER CORPORATION
CX50001
76635-574EA 547.83 CAD
76635-574
InTemp® CX Gateway
Data Loggers
The CX gateway works with wi-fi or ethernet communications to seamlessly link your InTemp CX data loggers to the InTempConnect cloud platform. The CX gateway automatically downloads data from in-range InTemp CX loggers and uploads it to the InTempConnect cloud platform, which sends automatic text/email notifications if alarm thresholds are exceeded.
  • Automates data download from all InTemp CX data loggers
  • Notifies you of temperature excursions in real time
  • Decreases risk of loss
  • Reduces time spent on data download and compliance