Solaris 4000 I Incubated Shakers
Supplier: Thermo Fisher Scientific
SK4001 SK4001CPKG SK4001MPKG
76633-006EA 16906 CAD
76633-006 76633-008 76633-010
Solaris 4000 I Incubated Shakers
Shakers Shaking Incubators
Solaris incubated orbital shakers feature Peltier chillers, units have a compact footprint, are designed to fit on the benchtop, user programmability for easy setup and reproducibility plus connectivity for easy monitoring.
  • Exceptional reliability with heavy-duty drive mechanism
  • Ability to create, download, and share up to 99 programs across multiple units
  • USB and ethernet connections for monitoring and data output
  • Refrigerant-free, environmentally friendly Peltier cooling, which uses 58% less energy compared to older models with traditional compressor
More Product Information