DNA/RNA Shield™ SafeCollect™ Swab Collection Kits
Supplier: Zymo Research
R1160 R1160-E R1161 R1161-E
76628-460EA 13.08 CAD
76628-460 76628-462 76628-454 76628-456
DNA/RNA Shield™ SafeCollect™ Swab Collection Kits
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Sputum Sample Collection
The DNA/RNA Shield SafeCollect swab collection kit is a user-friendly collection kit for stabilizing the nucleic acid content of samples collected with a swab. DNA/RNA shield completely inactivates harmful pathogens in the sample, and stabilizes the RNA in a sample for up to 30 days at room temperature. The safe collect design eliminates the risk of spilling the stabilization reagent during use, and is ideal for applications that require at-home self collection.
  • User-friendly, spill-free design allows for safe, convenient collection of swab samples
More Product Information