VWR® UHP Single Channel Pipettes and Pipette Sets
Supplier: VWR International
76627-784 76627-786 76627-802 76627-796 76627-772 76627-790 76627-776 76627-766 76627-792 76627-798 76627-770
76627-784EA 942.32 CAD
76627-784 76627-786 76627-802 76627-796 76627-772 76627-790 76627-776 76627-766 76627-792 76627-798 76627-770
VWR® UHP Single Channel Pipettes and Pipette Sets
Pipettes Micropipettes
The VWR® UHP single channel pipette is available in sizes from 2 to 1000 µl with a lightweight and durable design. These pipettes will provide you with low pipetting forces and the highest levels of accuracy and precision.
  • Autolocking volume mechanism to avoid volume drift while pipetting
  • Fully autoclaveable and UV resistant
  • Color coded ejector button and calibration switch make for easy identification
  • Universal fit with most common brands of pipette tips
  • Lightweight design, quality materials, low pipetting forces, and a 5-year warranty
These singleMore Product Information