Heptyl-β-D-thioglucopyranoside ≥97%

Supplier: AFG Bioscience

C54452-10G C54452-2G C54452-5G
76620-104EA 1740.65 CAD
76620-104 76620-100 76620-102
Heptyl-β-D-thioglucopyranoside ≥97%
Heptyl β-D-thioglucopyranoside
Formula: C₁₃H₂₆O₅S
Storage Temperature: Freezer
MDL Number: MFCD00043236
CAS Number: 85618-20-8

Order Now

Specification Test Results

Purity Limit > 97%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR