2,​3,​4,​6-​Tetra-​O-​acetyl-​D-​mannopyranose 95%

Supplier: AFG Bioscience

C53955-1G C53955-5G
76620-002EA 276.93 CAD
76620-002 76620-004
2,​3,​4,​6-​Tetra-​O-​acetyl-​D-​mannopyranose 95%
2,​3,​4,​6-​Tetra-​O-​acetyl-​D-​mannopyranose
Formula: C₁₄H₂₀O₁₀
Storage Temperature: Refrigerator
MDL Number: MFCD04039684
CAS Number: 140147-37-1

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR