α-Man-PEG4-Azide 95%

Supplier: AFG Bioscience

C51623-100MG C51623-50MG
76619-644EA 870.33 CAD
76619-644 76619-642
α-Man-PEG4-Azide 95%
α-Man-PEG4-Azide
Formula: C₁₄H₂₇N₃O₉
Storage Temperature: Freezer
CAS Number: 1632372-86-1

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR