2-​Acetamido-​2-​deoxy-​3,​4,​6-​tri-​O-​acetyl-​β-​D-​galactopyranosyl azide ≥95%

Supplier: AFG Bioscience

C51571-1G C51571-250MG C51571-500MG
76619-604EA 1503.29 CAD
76619-604 76619-600 76619-602
2-​Acetamido-​2-​deoxy-​3,​4,​6-​tri-​O-​acetyl-​β-​D-​galactopyranosyl azide ≥95%
2-​Acetamido-​2-​deoxy-​3,​4,​6-​tri-​O-​acetyl-​β-​D-​galactopyranosyl azide
Formula: C₁₄H₂₀N₄O₈
Storage Temperature: Refrigerator
CAS Number: 39541-20-3

Order Now

Specification Test Results

Purity Limit > 95%
Appearance Off-White Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR