Phenyl 2,3,4,6-tetra-O-acetyl-α-D-thiomannopyranoside 97%

Supplier: AFG Bioscience

C50717-10G C50717-1G C50717-5G
76619-200EA 1168.24 CAD
76619-200 76619-196 76619-198
Phenyl 2,3,4,6-tetra-O-acetyl-α-D-thiomannopyranoside 97%
Phenyl 2,3,4,6-tetra-O-acetyl-α-D-thiomannopyranoside
Formula: C₂₀H₂₄O₉S
Storage Temperature: Refrigerator
MDL Number: MFCD01862644
CAS Number: 13992-16-0

Order Now

Specification Test Results

Purity Limit 97%
Appearance white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR