Gamma Irradiated Sterility Maintenance Covers, Self-Seal, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
3312ST 3315ST 3315ST5 3320ST 3322ST 3330ST 3334ST 3335ST 3320ST3 3322ST3 3334ST3
76595-188CS 3435.35 CAD
76595-188 76595-180 76595-184 76595-176 76595-204 76595-190 76595-202 76595-200 76595-192 76635-470 76635-472 76635-474
Gamma Irradiated Sterility Maintenance Covers, Self-Seal, Keystone Cleanroom Products
Bags Sterilization Pouches
Ideal for pharmaceutical and biotech production applications.
  • Wide range of sizes
  • Convenient self-seal closure; no need for the use of or validation of a heat sealer
  • No tools required to open
  • Impervious to liquid sanitization agents
  • Visibility inside the bag allows for identification
  • Prolong the storage and shelf-life of products sterilized ‘in-house’
After unloading the autoclave, under Grade A LAF, place wrapped, steam sterilized items into sterility maintenance bags and seal closed. This technique captures the conditions of Grade A and protectsMore Product Information