Swann Morton Surgical Blade Handle #6, Ergonomic, Cincinnati Surgical
Supplier: CINCINNATI SURGICAL CO.
07SM6B-E
76583-016EA 71.27 CAD
76583-016
Swann Morton Surgical Blade Handle #6, Ergonomic, Cincinnati Surgical
Scalpels and Scalpel Assemblies Scalpel Handles
Fits size 20 to 27 blades.
  • Autoclavable