Sol-Care® Safety Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA
SN1815 SN2015 SN2110 SN2115 SN2210 SN2215 SN2310 SN2315 SN2510 SN2515 SN2558 SN2712 SN3012 SN1815B SN2115B SN2215B SN2510B SN2515B SN2558B
76582-908BX 39.59 CAD
76582-908 76582-910 76582-912 76582-914 76582-916 76582-918 76582-920 76582-922 76582-924 76582-926 76582-928 76582-930 76582-932 76645-704 76645-710 76645-714 76645-720 76645-722 76645-724
Sol-Care® Safety Needles
Syringe Needles
The Sol-Care safety needle is designed to help prevent needlestick injuries. Every needle is equipped with a pivoting safety shield mechanism that locks in place to lower the risk of accidental injury.
  • Two options for protective arm activation: Hard surface activation and thumb activation
  • Simple one-handed safety shield mechanism; audible click indicates safety mechanism is secure and locked in place
  • Protective arm does not obstruct visualization of injection site
  • Bevel-up needle allows for low-angle injections
Hubs, protective shields and packagingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable