VWR® Standard Laboratory Chest Freezer with Manual Defrost (15 cu. ft.)
Supplier: VWR International
76580-202
76580-202EA 3356.64 CAD
76580-202
VWR® Standard Laboratory Chest Freezer with Manual Defrost (15 cu. ft.)
Freezers Laboratory / General Purpose Freezers
These VWR® Standard Series manual defrost chest freezers provide quality cold storage at an economical pricepoint.
  • Mechanical thermostat
  • Keyed door lock
  • High density foam insulation
  • Two removable baskets
  • Manual defrost
These manual defrost laboratory chest freezers offer professional cold storage for scientific and industrial settings. Units come equipped with temperature alarms, keyed door locks, forced draft circulation, and precise temperature monitoring through microprocessor control. The Standard line provides multi-functional features in a cost-More Product Information