VWR® Plus Speciality Laboratory Refrigerators with Glass Door
Supplier: Horizon Scientific
VWR-HC-19 76580-156 76580-160 76580-164
76580-152EA 12376.33 CAD
76580-152 76580-156 76580-160 76580-164
VWR® Plus Speciality Laboratory Refrigerators with Glass Door
Refrigerators
These VWR® Plus Series laboratory refrigerators offer premium cold storage with superior temperature control.
  • Microprocessor temperature controller
  • Digital temperature display and alarm module
  • Audible, visual, senor error, and power failure alarms
  • Alarm validation and alarm mute with ring back
  • Keyed door lock
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These speciality glass door laboratory refrigerators offer professional cold storage for scientific and industrial settings. Units come equipped with temperature alarms, keyed door locks,More Product Information