Human Interleukin-1α (IL-1α) ELISA Kit, AFG Bioscience

Supplier: AFG Bioscience

EK710259
76578-348EA 646.43 CAD
76578-348
Human Interleukin-1α (IL-1α) ELISA Kit, AFG Bioscience
Assays ELISAs

  • Assay principle: Quantitative
  • Research topic: Immunology
  • Abbreviation: IL-1α
  • Detection wavelength: 450 nm

Order Now

SPECIFICATIONS

Environmentally Preferable
Target protein Interleukin-1α
Primary antibody reactivity
Assay Type Sandwich
Host Human
Sensitivity 0.5 pg/ml
Detection range 1.6 - 100 pg/ml
Sample type Serum, plasma, urine, tissue homogenates, cell culture supernates
Sample volume 50 - 100 µl
Time to Results 1 - 5 h
Regulatory status RUO
Storage temperature 2…8 °C
Shelf life 6 months (4 °C); 12 months (–20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR