CO₂ Incubators with Hot Air Sterilization, CB Series, Binder
Supplier: Binder
9640-0028 9640-0007 9640-0011 9640-0012 9640-0015 9640-0016
76575-504EA 22692.75 CAD
76575-504 76575-482 76575-484 76575-486 76575-488 76575-490 76575-492
CO₂ Incubators with Hot Air Sterilization, CB Series, Binder
Incubators CO₂ Incubators
CO₂ incubators with a built in CO₂ sensor which can be sterilized. Units offer optimum growth conditions for cell cultures and meet demanding requirements for best possible cell growth and maximum sample safety. The ANTI.PLENUM design minimizes surfaces due to the seamless, deep-drawn inner chamber without fixtures and featuring an integrated shelf support system. This reduces the risk of contamination, giving less surface area and fewer parts to clean manually, and removes the running costs of consumables. Reliable decontamination is provided by the standard-compliant 180 °C hot airMore Product Information