VWR® Aluminum Canisters and Stainless Steel Frames for Blood Bag Storage
Supplier: CRYSTAL TECHNOLOGY & INDUSTRIES
F-C-250-5 F-C-500-3 F-C-500-4 F-C-500-5 F-C-50-7 F-C-50-8 F-C-50-9 F-C-750-3 F-C-750-4 F-C-750-5 C-1000 C-250 C-50 C-500 C-750 F-C-1000-3 F-C-1000-4 F-C-1000-5 F-C-250-3 F-C-250-4
76559-078EA 74.87 CAD
76559-078 76559-086 76559-076 76559-082 76559-062 76559-088 76559-092 76559-084 76559-098 76559-060 76559-068 76559-064 76559-072 76559-094 76559-096 76559-066 76559-074 76559-090 76559-080 76559-070
VWR® Aluminum Canisters and Stainless Steel Frames for Blood Bag Storage
Boxes Storage Boxes
Organize, maximize storage and protect sample integrity of blood bags, bone marrow, stem cells, cord blood and plasma bags with durable canisters and frames.
  • Canisters designed to hold Baxter Fenwal, CharterMed, Gambro, Stericon, MedSep and origen blood bags
  • Multiple canister capacity options fit your storage needs
  • Canisters have locking handle and thumb hole for easy handling
  • Frames constructed of durable corrosion resistant stainless steel
  • Multiple frames options available to fit your storage requirements
  • Frames feature handles for easy retrieval of
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable