RheoPacs, Port Tops, Rheo Flexibles (Division of Rheo Engineering)
Supplier: RHEO ENGINEERING LLC
4002703 4002705 4002707 4002709 4002711 4002653 4002655 4002657 4002659 4002661 4002663 4002665 4002639 4002641 4002723 4002725 4002727 4002683 4002685 4002687 4002689 4002691 4002713 4002715 4002717 4002719 4002721 4002693 4002695 4002697 4002699 4002701 4002673 4002675 4002677 4002679 4002681 4002643 4002645 4002647 4002649 4002651 40026670 4002669 4002671
76557-618BX 562.62 CAD
76557-618 76557-624 76557-740 76557-532 76557-764 76557-530 76557-572 76557-732 76557-700 76557-702 76557-574 76557-600 76557-680 76557-692 76557-646 76557-566 76557-654 76557-770 76557-662 76557-688 76557-650 76557-720 76557-716 76557-534 76557-772 76557-718 76557-768 76557-644 76557-690 76557-670 76557-548 76557-674 76557-538 76557-608 76557-774 76557-564 76557-752 76557-760 76557-682 76557-684 76557-570 76557-706 76557-580 76557-636 76557-542
RheoPacs, Port Tops, Rheo Flexibles (Division of Rheo Engineering)
Mailers and Shippers
Rheo Flexibles provides a wide range of single-use transfer containers. The RheoPac family of products offers an excellent alternative to stainless intermediate transfer containers, and can be configured with many volumes and port options.
  • Constructed from RheoFlex 302 polyethylene material
  • FDA approved for packaging
  • USP and EP compliant
  • 21 CFR 177.1520 compliant
  • Matte finish
  • Anti-static film
Designed with integrated HDPE ports and handling loops to allow easy filling, transporting, and unloading.
Certifications: USP <661.1>, USP <661.2More Product Information