RheoPacs, Closed Tops, Rheo Flexibles (Division of Rheo Engineering)
Supplier: RHEO ENGINEERING LLC
4002598 4002599 4002600 4002601 4002611 4002581 40026120 4002613 4002614 4002615 4002616 4002629 4002630 4002631 4002617 4002625 4002626 4002627 4002628 4002582 4002583 4002584 4002585 4002586 4002593 4002594 4002595 4002596 4002597
76557-762BX 6285.24 CAD
76557-762 76557-696 76557-672 76557-660 76557-628 76557-704 76557-694 76557-736 76557-558 76557-632 76557-598 76557-648 76557-742 76557-604 76557-750 76557-638 76557-712 76557-710 76557-738 76557-634 76557-592 76557-540 76557-668 76557-616 76557-708 76557-748 76557-528 76557-734 76557-602
RheoPacs, Closed Tops, Rheo Flexibles (Division of Rheo Engineering)
Mailers and Shippers
Rheo Flexibles provides a wide range of single-use transfer containers. The RheoPac family of products offers an excellent alternative to stainless intermediate transfer containers, and can be configured with many volumes and port options.
  • Constructed from RheoFlex 302 polyethylene material
  • FDA approved for packaging
  • USP and EP compliant
  • 21 CFR 177.1520 compliant
  • Matte finish
  • Anti-static film
Designed with integrated HDPE ports and handling loops to allow easy filling, transporting, and unloading.
Certifications: USP <661.1>, USP <661.2More Product Information