Orion™ Lab Star PH111 pH/mV Bench Meters, Thermo Scientific

Supplier: Thermo Fisher Scientific
LSTAR1110 LSTAR1118 LSTAR1116 LSTAR1117 LSTAR1115
76557-402EA 952 CAD
76557-402 76557-408 76557-412 76557-416 76557-424
Orion™ Lab Star PH111 pH/mV Bench Meters, Thermo Scientific
Electrochemistry Meters pH and Multi-Parameter Meters
Measure pH or ORP simultaneously with mV and temperature using the Orion™ Lab Star PH111 pH/mV bench meter. To enable comprehensive measurement capabilities, measure pH in a range of -2.000 to 18.000 pH, measure ORP in a range of -2000.0 to +2000.0 mV and measure temperature in a range of -5.0 to 105.0 °C or 23.0 to 221.0 °F.

  • Budget-friendly pH meter for simplified pH testing and ORP testing
  • Large 5” backlit display makes viewing measurements easy
  • Small meter footprint saves valuable bench space in your lab
  • Date and time stamped logs support GLP compliance
More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
pH range –2.000...18.000 pH
pH resolution 0.1, 0.01, 0.001 pH
pH accuracy ±0.005 pH ±1 LSD (relative)
pH calibration 1 - 5 points
mV range ±2000 mV (mV/ORP)
mV resolution 0.1 mV (mV/ORP)
mV accuracy ±0.2 mV or ±0.05 % of the reading, whichever is greater (mV/ORP, relative)
mV calibration 1 point (mV/ORP)
Temperature range −5.0...105.0 °C (23.0...221.0 °F)
Temperature resolution 0.1 °C, 0.1 °F
Temperature accuracy ±0.3 °C, ±0.5 °F (relative)
Temperature calibration 1 point
Display 5" backlit LCD
Power supply 100 - 240 VAC, 50 - 60 Hz, 9 DC adapter, 1.3 A
Data logging - log function 500 sets with time and date
Memory Non-volatile
IP rating IP-54
Outputs Computer or printer
Input BNC, ATC
Alarm Yes, 1 - 168 hours or off (calibration alarm)
Calibration Automatic buffer recognition with manual buffer entry option
W×D×H 155.2×198.3×61.7 mm
Weight 700 g (1.54 lbs.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR