Orion™ Lab Star PH111 pH/mV Bench Meters, Thermo Scientific
Supplier: Thermo Fisher Scientific
LSTAR1110 LSTAR1118 LSTAR1116 LSTAR1117 LSTAR1115
76557-402EA 952 CAD
76557-402 76557-408 76557-412 76557-416 76557-424
Orion™ Lab Star PH111 pH/mV Bench Meters, Thermo Scientific
Electrochemistry Meters pH and Multi-Parameter Meters
Measure pH or ORP simultaneously with mV and temperature using the Orion™ Lab Star PH111 pH/mV bench meter. To enable comprehensive measurement capabilities, measure pH in a range of -2.000 to 18.000 pH, measure ORP in a range of -2000.0 to +2000.0 mV and measure temperature in a range of -5.0 to 105.0 °C or 23.0 to 221.0 °F.
  • Budget-friendly pH meter for simplified pH testing and ORP testing
  • Large 5” backlit display makes viewing measurements easy
  • Small meter footprint saves valuable bench space in your lab
  • Date and time stamped logs support GLP compliance
More Product Information